Anti-MEC (CCL28) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P297-50UG 500-P297-100UG 500-P297-1MG
10777-558EA 339.8 USD
10777-558 10777-560 10777-562
Anti-MEC (CCL28) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MEC (CCL28). Anti­Human MEC (CCL28)­specific antibody was purified by affinity chromatography employing an immobilized Human MEC matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MEC (CCL28)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR