Anti-IL-5 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P25-50UG 500-P25-100UG 500-P25-1MG
10771-236EA 354.54 USD
10771-236 10771-238 10781-472
Anti-IL-5 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-5. Anti­Human IL-5­specific antibody was purified by affinity chromatography employing an immobilized Human IL-5 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR