Anti-CXCL16 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P200-50UG 500-P200-100UG 500-P200-1MG
10776-830EA 339.8 USD
10776-830 10776-832 10776-834
Anti-CXCL16 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human CXCL16. Anti­Human CXCL16­specific antibody was purified by affinity chromatography employing an immobilized Human CXCL16 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: CXCL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR