Anti-IFN-gamma Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P122GBT-25UG 500-P122GBT-50UG 500-P122GBT-1MG
10776-172EA 339.8 USD
10776-172 10776-174 10776-176
Anti-IFN-gamma Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Rat IFN-γ. Anti­Rat IFN-γ­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IFN-gamma
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR