Anti-M-CSF Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P62GBT-25UG 500-P62GBT-50UG
10781-522EA 339.8 USD
10781-522 10781-524 10781-526
Anti-M-CSF Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine M-CSF. Anti­Murine M-CSF­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: M-CSF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR