Anti-4-1BB Receptor Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P167GBT-25UG 500-P167GBT-50UG
10771-454EA 354.54 USD
10771-454 10776-546 10776-548
Anti-4-1BB Receptor Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human 4-1BB Receptor. Anti­Human 4-1BB Receptor­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: 4-1BB Receptor
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR