Anti-SDF-1alpha (CXCL12) Goat Polyclonal Antibody

Supplier: Peprotech
500-P164G-50UG 500-P164G-100UG 500-P164G-1MG
10776-522EA 354.54 USD
10776-522 10776-524 10776-526
Anti-SDF-1alpha (CXCL12) Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine SDF-1α (CXCL12). Anti­Murine SDF-1α (CXCL12)­specific antibody was purified by affinity chromatography employing an immobilized Murine SDF-1α matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: SDF-1alpha (CXCL12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR