Murine TNF-α Standard TMB ELISA Development Kit, PeproTech
Supplier: Peprotech
900-T54 900-TM54
10771-676KT 623.13 USD
10771-676 10777-932
Murine TNF-α Standard TMB ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Murine TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant mTNF-α in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant mTNF-α in a sandwich ELISA format. Using the ELISA protocol the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay mTNF-α in approximately 1000 ELISA plate wells.