Human SDF-1α (CXCL12) Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K92 900-M92
10771-542KT 643.41 USD
10771-542 10777-842
Human SDF-1α (CXCL12) Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Human SDF-1α (CXCL12) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Human SDF-1α (CXCL12) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human SDF-1α (CXCL12) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Human SDF-1α (CXCL12) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay Human SDF-1α (CXCL12) in approximately 1000 ELISA plate wells.