Rat IL-6 Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K86 900-M86
10771-534KT 577.11 USD
10771-534 10777-836
Rat IL-6 Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Rat IL-6 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat IL-6 in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Rat IL-6 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat IL-6 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay rat IL-6 in approximately 1000 ELISA plate wells.