3-Aminomethyltetrahydrofuran ≥98.8%

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

02180-1 02180-2 02180-3
103536-056EA 23.08 USD
103536-056 103536-058 103536-060
3-Aminomethyltetrahydrofuran ≥98.8%
3-Aminomethyltetrahydrofuran
Formula: C₅H₁₁NO
MW: 101.149 g/mol
MDL Number: MFCD08234925
CAS Number: 165253-31-6

Order Now

Specification Test Results

Purety 98.8.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR