(R)-3-(Boc-amino)piperidine ≥97%

Synonyms: tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate

00803-1 00803-2 00803-3
103534-480EA 30.77 USD
103534-480 103534-482 103534-484
(R)-3-(Boc-amino)piperidine ≥97%
(R)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03093382
CAS Number: 309956-78-3

Order Now

Specification Test Results

Purety 97%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR