3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester ≥99.3%

Synonyms: Methyl 3-Bromopyrrole-2-carboxylate

ZJ0001-1 ZJ0001-2 ZJ0001-3
103536-726EA 92.31 USD
103536-726 103536-728 103536-730
3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester ≥99.3%
3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester
Formula: C₆H₆BrNO₂
MW: 204.03 g/mol
MDL Number: MFCD23106341
CAS Number: 941714-57-4

Order Now

Specification Test Results

Purety 99.3%
Appearance Off-white crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR