4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥99.0%

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Y2010257-1 Y2010257-2 Y2010257-3
103536-720EA 32 USD
103536-720 103536-722 103536-724
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥99.0%
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₅H₄N₅I
MW: 261.03 g/mol
MDL Number: MFCD03787931
CAS Number: 151266-23-8

Order Now

Specification Test Results

Purety 99.0%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR