N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥90.0%

Synonyms: Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine

Y2010235-1 Y2010235-2 Y2010235-3
103536-714EA 12.31 USD
103536-714 103536-716 103536-718
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥90.0%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Formula: C₁₃H₂₃NOSi
MW: 237.42 g/mol
MDL Number: MFCD00674005
CAS Number: 93102-05-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Purety 90.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR