Methyl-1H-indazole-3-carboxylate ≥97.6%

Synonyms: Methyl 1H-indazole-3-carboxylate

Y2010211-1 Y2010211-2 Y2010211-3
103536-708EA 18.46 USD
103536-708 103536-710 103536-712
Methyl-1H-indazole-3-carboxylate ≥97.6%
Methyl 1H-indazole-3-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
MDL Number: MFCD01138133
CAS Number: 43120-28-1

Order Now

Specification Test Results

Purety 97.6%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR