3-Amino-2-chloropyridine ≥97.0%

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

Y2010190-1 Y2010190-2 Y2010190-3
103536-702EA 12.31 USD
103536-702 103536-704 103536-706
3-Amino-2-chloropyridine ≥97.0%
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD00006238
CAS Number: 6298-19-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Orange powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR