6-Oxospiro[3.3]heptane-2-carboxylic acid ≥95.1%

Y2010183-1 Y2010183-2 Y2010183-3
103536-690EA 153.85 USD
103536-690 103536-692 103536-694
6-Oxospiro[3.3]heptane-2-carboxylic acid ≥95.1%
6-Oxospiro[3.3]heptane-2-carboxylic acid
Formula: C₈H₁₀O₃
MDL Number: MFCD08436470
CAS Number: 889944-57-4

Order Now

Specification Test Results

Purety 95.1%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR