(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥97.0%

Synonyms: tert-Butyl (6-oxospiro[3.3]heptan-2-yl)carbamate, tert-butyl N-{6-oxospiro[3.3]heptan-2-yl}carbamate

Y2010182-1 Y2010182-2 Y2010182-3
103536-684EA 230.77 USD
103536-684 103536-686 103536-688
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥97.0%
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Formula: C₁₂H₁₉NO₃
MW: 225.29 g/mol
MDL Number: MFCD11858160
CAS Number: 1118786-86-9

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR