cis-tert-Butyl-3-hydroxycyclobutylcarbamate ≥97.0%

Synonyms: tert-Butyl (cis-3-hydroxycyclobutyl)carbamate

Y2010162-1 Y2010162-2 Y2010162-3
103535-602EA 76.92 USD
103535-602 103535-604 103535-606
cis-tert-Butyl-3-hydroxycyclobutylcarbamate ≥97.0%
cis-tert-Butyl-3-hydroxycyclobutylcarbamate
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD09038208
CAS Number: 389890-43-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR