1-Boc-3-fluoro-4-piperidone ≥98.9%

Synonyms: 1-Piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, tert-Butyl 3-Fluoro-4-oxo-1-piperidinecarboxylate, tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate, 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, 3-Fluoro-4-oxo-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-fluoro-4-piperidone

Y2010157-1 Y2010157-2 Y2010157-3
103535-596EA 46.15 USD
103535-596 103535-598 103535-600
1-Boc-3-fluoro-4-piperidone ≥98.9%
1-Boc-3-fluoro-4-piperidone
Formula: C₁₀H₁₆FNO₃
MW: 217.24 g/mol
MDL Number: MFCD10566554
CAS Number: 211108-50-8

Order Now

Specification Test Results

Purety 98.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR