5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥97.0%

Y2010140-1 Y2010140-2 Y2010140-3
103535-590EA 138.46 USD
103535-590 103535-592 103535-594
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥97.0%
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₃BrClN₃
MW: 232.4700 g/mol
MDL Number: MFCD09909707
CAS Number: 1060816-58-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR