7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1) ≥97.0%

Synonyms: 7-Oxa-2-azaspiro[3.5]nonane oxalate(2:1), 7-oxa-2-azaspiro[3.5]nonane hemioxalate

Y2010131-01-3 Y2010131-01-1 Y2010131-01-2
103535-588EA 553.85 USD
103535-588 103536-678 103536-680
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1) ≥97.0%
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)
Formula: C₁₆H₂₈N₂O₆
MW: 344.41 g/mol
MDL Number: MFCD22422285
CAS Number: 1523571-04-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR