2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) ≥97.0%

Synonyms: 2-Oxa-6-azaspiro[3.3]heptane hemioxalate

Y2010010-01-1 Y2010010-01-2 Y2010010-01-3
103536-672EA 61.54 USD
103536-672 103536-674 103536-676
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) ≥97.0%
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1)
Formula: C₁₂H₂₀N₂O₆
MW: 144.15 g/mol
MDL Number: MFCD16652332
CAS Number: 1045709-32-7
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR