4-Iodo-1-methyl-1H-pyrazole ≥99.5%

XA8067-1 XA8067-2 XA8067-3
103536-660EA 8.62 USD
103536-660 103536-662 103536-664
4-Iodo-1-methyl-1H-pyrazole ≥99.5%
4-Iodo-1-methyl-1H-pyrazole
Formula: C₄H₅IN₂
MW: 208.0000 g/mol
MDL Number: MFCD05663860
CAS Number: 39806-90-1

Order Now

Specification Test Results

Purety 99.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR