Boc-D-Pro-OH ≥97.0%

Synonyms: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

XA1131-1 XA1131-2 XA1131-3
103536-648EA 30.77 USD
103536-648 103536-650 103536-652
Boc-D-Pro-OH ≥97.0%
Boc-D-Pro-OH
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD00063226
CAS Number: 37784-17-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR