3,3-Bis(chloromethyl)oxetane ≥95.0%

W2011001-1 W2011001-2 W2011001-3
103536-642EA 30.77 USD
103536-642 103536-644 103536-646
3,3-Bis(chloromethyl)oxetane ≥95.0%
3,3-Bis(chloromethyl)oxetane
Formula: C₅H₈Cl₂O
MDL Number: MFCD00027377
CAS Number: 78-71-7

Order Now

Specification Test Results

Purety 95.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR