tert-Butyl 4-[3-(methanesulfonyloxy)propyl]piperazine-1-carboxylate hydrochloride ≥97%

U12235-01-1 U12235-01-2 U12235-01-3
103536-636EA 12.31 USD
103536-636 103536-638 103536-640
tert-Butyl 4-[3-(methanesulfonyloxy)propyl]piperazine-1-carboxylate hydrochloride ≥97%
tert-Butyl 4-[3-(methanesulfonyloxy)propyl]piperazine-1-carboxylate hydrochloride
Formula: C₁₃H₂₆N₂O₅S·HCl
MW: 358.89 g/mol
CAS Number: 1630906-98-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purety 97%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR