(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine ≥99.8%

Synonyms: PSI-6206, 1-[(2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

SQ8028-1 SQ8028-2 SQ8028-3
103536-624EA 30.77 USD
103536-624 103536-626 103536-628
(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine ≥99.8%
(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine
Formula: C₁₀H₁₃FN₂O₅
MW: 260.219 g/mol
MDL Number: MFCD27918669
CAS Number: 863329-66-2

Order Now

Specification Test Results

Purety 99.8%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR