N-(Ethoxycarbonyl)nortropinone ≥99.3%

Synonyms: Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone

Q11251-1 Q11251-2 Q11251-3
103536-618EA 138.46 USD
103536-618 103536-620 103536-622
N-(Ethoxycarbonyl)nortropinone ≥99.3%
N-(Ethoxycarbonyl)nortropinone
Formula: C₁₀H₁₅NO₃
MW: 197.23 g/mol
MDL Number: MFCD00078171
CAS Number: 32499-64-2

Order Now

Specification Test Results

Purety 99.3%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR