Methyl 5-amino-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥97%

N20122116-1 N20122116-2 N20122116-3
103536-610EA 230.77 USD
103536-610 103536-612 103536-614
Methyl 5-amino-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥97%
Methyl 5-amino-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Formula: C₉H₉N₃O₂
CAS Number: 952182-18-2

Order Now

Specification Test Results

Purety 97%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR