tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate ≥88.8%

N20122103-1 N20122103-2 N20122103-3
103536-604EA 115.38 USD
103536-604 103536-606 103536-608
tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate ≥88.8%
tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate
Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD12068393
CAS Number: 937046-96-3

Order Now

Specification Test Results

Purety 88.8%
Appearance Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR