3-Aminopiperidine-2,6-dione hydrochloride ≥97.0%

Synonyms: 3-aminopiperidine-2,6-dione hydrochloride, 2,6-Dioxopiperidine-3-ammonium chloride

N20122102-1-1 N20122102-1-2 N20122102-1-3
103536-598EA 276.92 USD
103536-598 103536-600 103536-602
3-Aminopiperidine-2,6-dione hydrochloride ≥97.0%
3-Aminopiperidine-2,6-dione hydrochloride
Formula: C5H9ClN2O2
MDL Number: MFCD11042437
CAS Number: 24666-56-6

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR