Cis-4-Hydroxy-D-proline hydrochloride ≥97.0%

Synonyms: (2R,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride, H-cis-D-Hyp-OH·HCl

N20122057-1-1 N20122057-1-2
103536-592EA 30.77 USD
103536-592 103536-594
Cis-4-Hydroxy-D-proline hydrochloride ≥97.0%
Cis-4-Hydroxy-D-proline hydrochloride
Formula: C₅H₁₀ClNO₃
MW: 167.5908 g/mol
MDL Number: MFCD09749867
CAS Number: 77449-94-6

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR