(R)-1-tert-Butyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxylate ≥97.2%

Synonyms: 1-tert-butyl 2-methyl (2R)-4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-Oxo-Pro-OMe, (R)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate

N20122054-1 N20122054-2 N20122054-3
103536-586EA 55.38 USD
103536-586 103536-588 103536-590
(R)-1-tert-Butyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxylate ≥97.2%
(R)-1-tert-Butyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₁₇NO₅
MW: 243.26 g/mol
MDL Number: MFCD09954936
CAS Number: 256487-77-1

Order Now

Specification Test Results

Purety 97.2%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR