(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate ≥98.5%

Synonyms: 1-tert-butyl 3-methyl (3S)-piperazine-1,3-dicarboxylate

N20121936-1 N20121936-2 N20121936-3
103536-580EA 46.15 USD
103536-580 103536-582 103536-584
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate ≥98.5%
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
Formula: C₁₁H₂₀N₂O₄
MW: 244.2875 g/mol
MDL Number: MFCD04115326
CAS Number: 314741-39-4

Order Now

Specification Test Results

Purety 98.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR