5-Hydroxypicolinic acid ≥99.0%

Synonyms: 5-hydroxypyridine-2-carboxylic acid

N20121773-1 N20121773-2 N20121773-3
103536-574EA 23.08 USD
103536-574 103536-576 103536-578
5-Hydroxypicolinic acid ≥99.0%
5-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00661309
CAS Number: 15069-92-8

Order Now

Specification Test Results

Purety 99.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR