4-Bromo-2-pyridone ≥97.8%

Synonyms: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

N20121749-1 N20121749-2 N20121749-3
103536-568EA 30.77 USD
103536-568 103536-570 103536-572
4-Bromo-2-pyridone ≥97.8%
4-Bromo-2-pyridone
Formula: C₅H₄BrNO
MW: 174.00 g/mol
MDL Number: MFCD00234041
CAS Number: 36953-37-4

Order Now

Specification Test Results

Purety 97.8%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR