(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate ≥97.8%

Synonyms: 1-tert-butyl 2-methyl (2S)-4,4-difluoropyrrolidine-1,2-dicarboxylate, (S)-1-tert-Butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate

N20121533-1 N20121533-2 N20121533-3
103536-562EA 76.92 USD
103536-562 103536-564 103536-566
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate ≥97.8%
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₁₇F₂NO₄
MW: 265.26 g/mol
MDL Number: MFCD03094918
CAS Number: 203866-17-5

Order Now

Specification Test Results

Purety 97.8%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR