trans-4-(Boc-amino)cyclohexanol ≥99.9%

Synonyms: tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected

N20121349-1 N20121349-2 N20121349-3
103536-556EA 30.77 USD
103536-556 103536-558 103536-560
trans-4-(Boc-amino)cyclohexanol ≥99.9%
trans-4-(Boc-amino)cyclohexanol
Formula: C₁₁H₂₁NO₃
MW: 215.28 g/mol
MDL Number: MFCD03844613
CAS Number: 111300-06-2

Order Now

Specification Test Results

Purety 99.9%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR