2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride ≥98.0%

N20121338-01-1 N20121338-01-2 N20121338-01-3
103536-550EA 100 USD
103536-550 103536-552 103536-554
2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride ≥98.0%
2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride
Formula: C₅H₁₀ClNO
MDL Number: MFCD07798642
CAS Number: 909186-56-7

Order Now

Specification Test Results

Purety 98.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR