3-Methyloxetan-3-ol ≥97.5%

Synonyms: 3-Hydroxy-3-methyloxetane

N20121278-1 N20121278-2 N20121278-3
103536-538EA 150.77 USD
103536-538 103536-540 103536-542
3-Methyloxetan-3-ol ≥97.5%
3-Methyloxetan-3-ol
Formula: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD18909291
CAS Number: 162816-08-2

Order Now

Specification Test Results

Purety 97.5%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR