cis-3-Hydroxycyclobutanecarboxylic acid ≥97.0%

N20121262-1 N20121262-2 N20121262-3
103536-532EA 115.38 USD
103536-532 103536-534 103536-536
cis-3-Hydroxycyclobutanecarboxylic acid ≥97.0%
cis-3-Hydroxycyclobutanecarboxylic acid
Formula: C₅H₈O₃
MW: 116.1200 g/mol
MDL Number: MFCD12964956
CAS Number: 552849-33-9

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR