6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol ≥97.0%

Synonyms: Ticagrelor Related Compound 1, (3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-ol, (3aR,4S,6R,6aS)-6-amino-2,2-dimethyl-hexahydrocyclopenta[d][1,3]dioxol-4-ol

N20121172-1 N20121172-2 N20121172-3
103536-514EA 30.77 USD
103536-514 103536-516 103536-518
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol ≥97.0%
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD20527303
CAS Number: 155899-66-4

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR