Methyl 1H-pyrazole-4-carboxylate ≥99.0%

N20121105-1 N20121105-2 N20121105-3
103536-508EA 38.46 USD
103536-508 103536-510 103536-512
Methyl 1H-pyrazole-4-carboxylate ≥99.0%
Methyl 1H-pyrazole-4-carboxylate
Formula: C₅H₆N₂O₂
MW: 126.113 g/mol
MDL Number: MFCD00297363
CAS Number: 51105-90-9

Order Now

Specification Test Results

Purety 99.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR