2,4-Dichloro-3-nitropyridine ≥99.7%

N20120972-1 N20120972-2 N20120972-3
103536-496EA 18.46 USD
103536-496 103536-498 103536-500
2,4-Dichloro-3-nitropyridine ≥99.7%
2,4-Dichloro-3-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
MW: 192.9876 g/mol
MDL Number: MFCD09261088
CAS Number: 5975-12-2

Order Now

Specification Test Results

Purety 99.7%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR