trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride ≥97%

Synonyms: trans-4-Fluoropyrrolidin-3-ol hydrochloride

N20120938-1-1 N20120938-1-2 N20120938-1-3
103535-586EA 461.54 USD
103535-586 103536-492 103536-494
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride ≥97%
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride
Formula: C₄H₈FNO·HCl
MW: 141.572 g/mol
MDL Number: MFCD23105899
CAS Number: 2006333-41-9

Order Now

Specification Test Results

Purety 97%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR