1-Boc-4-cyanopiperidine ≥97.0%

Synonyms: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

N20120903-1 N20120903-2 N20120903-3
103535-580EA 33.23 USD
103535-580 103535-582 103535-584
1-Boc-4-cyanopiperidine ≥97.0%
1-Boc-4-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
MDL Number: MFCD01861223
CAS Number: 91419-52-2

Order Now

Specification Test Results

Purety 97.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR