3-Amino-5-methyl-1H-pyrazole ≥97.0%

Synonyms: 3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine

N20120842-1 N20120842-2 N20120842-3
103535-574 103535-576 103535-578
3-Amino-5-methyl-1H-pyrazole ≥97.0%
3-Amino-5-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR