(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥98.1%

Synonyms: Boc-Cis-4-F-Pro-OH, N-Boc-cis-4-fluoro-L-proline, N-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid, (2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid

N20120717-1 N20120717-2 N20120717-3
103536-484EA 30.77 USD
103536-484 103536-486 103536-488
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥98.1%
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
MDL Number: MFCD04973957
CAS Number: 203866-13-1

Order Now

Specification Test Results

Purety 98.1%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR